missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide in ethanol, BAKER ANALYZED™, J.T. Baker™
4417.50 NOK valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
1 L
Description
A light red color can occur in time due to product characteristics
Specifications
CAS | 1310-58-3 |
Molecular Formula | HKO |
Concentration | 0.5 mol/L (0.5 N) |
MDL Number | MFCD00003553 |
Synonym | Caustic potash |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Quantity | 1 L |
SMILES | [OH-].[K+] |
IUPAC Name | potassium hydroxide |
Molecular Weight (g/mol) | 56.11 |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'