missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide (~85%), TRC

Product Code. 30007110
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007110 500 g 500g
30003866 250 g 250g
30008424 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007110 Supplier Toronto Research Chemicals Supplier No. TRCP695060500G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula H K O
Molecular Weight (g/mol) 56.11
Synonym Potassium hydroxide
IUPAC Name hydroxypotassium
SMILES O[K]

Specifications

InChI Formula InChI=1S/K.H2O/h;1H2/q+1;/p-1
Quantity 500 g
Formula Weight 55.9664
Chemical Name or Material Potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.