Learn More
Potassium hydroxide, 0.5N Standardized Solution
Product Code. p-4407743
Click to view available options
Quantity:
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
Product Code. 11393697
Brand: Thermo Scientific Alfa Aesar 035595.K2
Request Bulk or Custom Format
CAS: 1310-58-3 | HKO | 56.11 g/mol
Potassium hydroxide, 0.5N Standardized Solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]
Specifications
Color Colorless
Physical Form Liquid
Quantity 1 L
Solubility Information Freely soluble in water. Soluble in ethanol.
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Appearance Colorless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Potassium hydroxide