missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogensulfate, Fused, ACS Reagent, Mixture of K2S2O7 and KHSO4, Honeywell Fluka™

Product Code. p-7120707 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15672230

Brand: Honeywell-Fluka 223697250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, Mixture of K2S2O7 and KHSO4

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7, 7790-62-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Color White
Quantity 250 g
MDL Number MFCD00011404
UN Number UN2509
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.512g/mL at 25°C
Chemical Name or Material Potassium Hydrogensulfate, Fused, Mixture of K2S2O7 and KHSO4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.