missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogensulfate, Fused, ACS Reagent, Mixture of K2S2O7 and KHSO4, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Color | White |
| Quantity | 1 kg |
| MDL Number | MFCD00066385 |
| UN Number | UN2509 |
| Grade | ACS Reagent |
| Packaging | Poly Bottle |
| Appearance | Powder |
| Density | 2.512g/mL at 25°C |
| Chemical Name or Material | Potassium Hydrogensulfate, Fused, Mixture of K2S2O7 and KHSO4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction