missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, for HPLC

Product Code. p-7214472 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 16514254

Brand: Thermo Scientific 463850500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
IUPAC Name potassium 2-carboxybenzoate
SMILES [K+].OC(=O)C1=CC=CC=C1C([O-])=O

Specifications

Melting Point 295.0°C to 300.0°C
Quantity 50 g
Linear Formula 2-(HO2C)C6H4CO2K
Solubility Information Solubility aq. soln.: 80 g/L (20°C). Other solubilities: 330 g/L water (100°C), slightly soluble in alcohol
Formula Weight 204.22
Grade HPLC
Chemical Name or Material Potassium hydrogen phthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.