Learn More
Potassium Hydrogen Monopersulfate, Sigma-Aldrich™
Product Code. 15810391
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810391 500 g 500g
15800391 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15810391 Supplier Sigma Aldrich Supplier No. 8184010500
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70693-62-8
Specifications
Quantity 500 g
MDL Number MFCD00040551
Synonym Potassium hydrogen monopersulfate, PMPS, KMPS, Potassium hydrogen monopersulfate
Molecular Weight (g/mol) 614.76
Grade Research
Density 2.35 g/cm3 at 20°C
Chemical Name or Material Potassium Hydrogen Monopersulfate