Learn More
Potassium hydrogen carbonate, Supelco™
Specifications
Specifications
| Chemical Name or Material | Potassium hydrogen carbonate |
| CAS | 298-14-6 |
| Quantity | 25 kg |
| Density | 2.17 g/cm³ at 20°C |
| Synonym | Potassium hydrogen carbonate, Potassium bicarbonate, Potassium hydrogen carbonate |
| Molecular Weight (g/mol) | 100.12 |
| Grade | Not Available |