Learn More
Potassium hexachloroosmate(IV), Os 38.7% min
Product Code. 11326568
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326568 5 g 5g
11306568 0.25 g 0.25g
11316568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11326568 Supplier Thermo Scientific Alfa Aesar Supplier No. 012177.06
Request Bulk or Custom Format
CAS: 16871-60-6
Potassium hexachloroosmate(IV) used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16871-60-6
MDL Number MFCD00011371
Specifications
Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 5 g
Linear Formula K2OsCl6
Merck Index 14,7635
Solubility Information Soluble in water; Sparingly soluble in alcohol
Formula Weight 481.12
Concentration or Composition (by Analyte or Components) Os 38.7% min.
Sensitivity Hygroscopic
Chemical Name or Material Potassium hexachloroosmate(IV)