Learn More
Potassium Cyanide, Supelco™
Product Code. 15874490
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874490 1 kg 1kg
15854490 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15874490 Supplier Supelco Supplier No. 1049671000
 Restricted Item
Estimated Shipment: 19-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151-50-8
Specifications
Boiling Point 1625°C/1013 hPa
Melting Point 634°C
Quantity 1 kg
MDL Number MFCD00011397
Synonym Potassium cyanide, Cyanogen potassium, Potassium cyanide
Molecular Weight (g/mol) 65.12
Grade Research
Density 1.55 g/cm3 at 20°C
Chemical Name or Material Potassium Cyanide