Learn More
Potassium Chloride Solution, EU Pharmacopoeia Grade, 0.5M, Fisher Chemical™
Product Code. 12933574 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. 12933574
Brand: Fisher Chemical VL 256 1000
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.5mol/L