missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 1100μS, Chem-Lab

Product Code. 11993083
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11993083 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11993083 Supplier Chem-Lab Supplier No. CL0311200500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calibrates conductivity measurement equipment. Potassium Chloride Conductivity Standard, 1100μS, Chem-Lab is precisely concentrated to a specific LF electrolyte conductivity level in order to calibrate and verify the performance of conductivity meters and probes.

Use of conductivity standard ensures that instruments are properly calibrated for accurate readings. By placing probes into a solution with an identified transfer ability, the equipment settings can be adjusted prior to testing. KCl solutions are used to calibrate conductivity instruments that read directly in microsiemens or millisiemens. Potassium chloride is a very stable salt and is an international calibration standard for conductivity measurement.

Specifications

CAS 7447-40-7
Odor Odorless
Density 1.00g/mL
Conductivity 1100μS at 25°C (±1%)
Physical Form Liquid
Molecular Formula ClK
MDL Number MFCD00011360
Color Colorless
pH ±5
For Use With (Equipment) Conductivity Meter
Solubility Soluble
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.