Learn More
Potassium Chloride Conductivity Standard, 24700μS, Chem-Lab
Product Code. 11986696
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11986696
Brand: Chem-Lab CL03.1124.0500
Calibrates conductivity measurement equipment. Potassium Chloride Conductivity Standard, 24700μS, Chem-Lab is precisely concentrated to a specific LF electrolyte conductivity level in order to calibrate and verify the performance of conductivity meters and probes.
Use of conductivity standard ensures that instruments are properly calibrated for accurate readings. By placing probes into a solution with an identified transfer ability, the equipment settings can be adjusted prior to testing. KCl solutions are used to calibrate conductivity instruments that read directly in microsiemens or millisiemens. Potassium chloride is a very stable salt and is an international calibration standard for conductivity measurement.
Specifications
CAS 7447-40-7
Odor Odorless
Density 1.01g/mL
Conductivity 24700μS at 25°C
22400μS at 20°C (±1%)
Physical Form Liquid
Molecular Formula ClK
Concentration 0.2 mol/L
MDL Number MFCD00011360
Color Colorless
pH ±5
For Use With (Equipment) Conductivity Meter
Solubility Soluble
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol