Learn More
Potassium Chloride, 3mol/l Solution, For Filling Electrodes, Scharlab
Product Code. 11344264
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344264 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11344264 Supplier Scharlab Supplier No. PO02050250
  • ONLINE EXCLUSIVE PRICE
    356.00 NOK
    327.73 NOK / 250mL
    Save 28.27 NOK
    7% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Used for filling electrodes in electroanalysis, in analytical chemistry and as a laboratory reagent.
  • Molecular Formula: KCl
  • Molar Mass: 74.56g/mol
  • CAS: 7447-40-7
  • EIN ECS No.: 231-211-8
Specifications
CAS 7447-40-7
Quantity 250 mL
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Density 1.13g/cm3