missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium carbonate, EMSURE™ MilliporeSigma™
Specifications
Specifications
| Melting Point | 891°C |
| CAS | 584-08-7 |
| Color | White |
| pH | 11.5 to 12.5 (50g/L, H2O, 20°C) |
| Physical Form | Solid |
| Quantity | 500 g |
| Molecular Formula | CK2O3 |
| MDL Number | MFCD00011382 |
| Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate |
| Solubility Information | 1120g/L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?