missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, puriss., Honeywell Fluka™
BIOPHARMA

Product Code. p-7071695 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
25kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15620560

Brand: Honeywell-Fluka 02110100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]

Specifications

CAS 7758-02-3, 14708-99-7
Quantity 100 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium bromide
Formula Weight 119g/mol
Percent Purity 99.5 to 100.5%
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Chemical Name or Material Potassium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.