Learn More
Potassium for AAS 1g/L HCl matrix, Fisher chemical™
BIOPHARMA
Product Code. 12923175 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
12923175 500 mL 500mL
12913175 100 mL 100mL
2 options
Product Code. 12923175
Brand: Fisher Chemical B 028 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 500 mL