Learn More
Potassium 5-(9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl)tetrazol-2-ide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 100299-08-9 |
| Molecular Formula | C10H7KN6O |
| MDL Number | MFCD01690051 |
| Quantity | 250 mg |
| SMILES | O=C1C(C2=N[N-]N=N2)=CN=C3N1C=CC=C3C.[K+] |
| Molecular Weight (g/mol) | 266.30 |
| Percent Purity | 98% |
| Chemical Name or Material | Potassium 5-(9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl)tetrazol-2-ide |