missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium 4-nitrophenyltrifluoroborate, 95%

Product Code. 15439967
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
15439967 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 15439967

Brand: Alfa Aesar™ H53208.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 850623-71-1
Melting Point 288°C to 290°C
Molecular Formula C6H4BF3KNO2
MDL Number MFCD04115766
Quantity 250 mg
Synonym potassium 4-nitrophenyl trifluoroborate, potassium trifluoro 4-nitrophenyl borate, potassium 4-nitrophenyltrifluoroborate, potassium trifluoro 4-nitrophenyl boranuide, potassium trifluoro 4-nitrophenyl borate 1-
InChI Key VUFAMHLXAXZYSD-UHFFFAOYSA-N
SMILES [B-](C1=CC=C(C=C1)[N+](=O)[O-])(F)(F)F.[K+]
IUPAC Name potassium;trifluoro-(4-nitrophenyl)boranuide
Molecular Weight (g/mol) 229.007
PubChem CID 44886958
Formula Weight 229.01
Percent Purity 95%
Chemical Name or Material Potassium 4-nitrophenyltrifluoroborate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.