Learn More
Potassium 4-carbamoylphenyltrifluoroborate, 96%, Thermo Scientific™
Product Code. 15429827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429827 5 g 5g
15419827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15429827 Supplier Thermo Scientific Alfa Aesar Supplier No. H53054.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850623-63-1
Molecular Formula C7H6BF3KNO
Molecular Weight (g/mol) 227.035
MDL Number MFCD04115758
InChI Key BMTORUMAAKNFNW-UHFFFAOYSA-N
Synonym potassium 4-aminocarbonylphenyl trifluoroborate, potassium 4-carbamoylphenyl trifluoroborate, potassium 4-carbamoylphenyltrifluoroborate, potassium 4-carbamoylphenyl trifluoroboranuide, potassium 4-carbamoylphenyl-trifluoroboranuide, potassium ion 4-carbamoylphenyl trifluoroboranuide, potassium 4-carbamoylphenyl trifluoro borate 1-, potassium 4-aminocarbonylphenyl-tris fluoranyl boranuide
PubChem CID 44717215
IUPAC Name potassium;(4-carbamoylphenyl)-trifluoroboranuide
SMILES [B-](C1=CC=C(C=C1)C(=O)N)(F)(F)F.[K+]
Specifications
Melting Point >300°C
Quantity 5 g
Formula Weight 227.03
Percent Purity 96%
Chemical Name or Material Potassium 4-carbamoylphenyltrifluoroborate