Learn More
Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 868-14-4 |
| Molecular Formula | C4H5KO6 |
| MDL Number | MFCD00065392 |
| Quantity | 100 g |
| SMILES | O=C([O-])[C@H](O)[C@@H](O)C(O)=O.[K+] |
| Molecular Weight (g/mol) | 188.18 |
| Percent Purity | 98% |
| Chemical Name or Material | Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate |