Learn More
Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 25102-12-9 |
| Molecular Formula | C10H18K2N2O10 |
| MDL Number | MFCD00150036 |
| Quantity | 25 g |
| SMILES | O=C([O-])CN(CC([O-])=O)CCN(CC(O)=O)CC(O)=O.[H]O[H].[H]O[H].[K+].[K+] |
| Molecular Weight (g/mol) | 404.45 |
| Percent Purity | 98% |
| Chemical Name or Material | Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate |