Learn More
Notrax Posture Mat™ Anti-Fatigue Mat for Sit-Stand Desks
Product Code. p-7227292
Change view
Click to view available options
Length (Metric):
70 cm
99 cm
Unit Size:
Each
Product Code. Length (Metric) unitSize
17702498 70 cm Each
17712498 99 cm Each
2 options
Product Code. 17702498
Brand: Notrax 426S7052GY
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Posture Mat™ Anti-Fatigue Mat for Sit-Stand Desks is an exceptionally soft and comfortable anti-fatigue mat for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations.
  • Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations
  • The rebound of the foam results in an energizing bounce effect that stimulates automatic balance checks to improve posture and constant micro movements to maintain blood flow
  • The ideal anti-fatigue mat for adjustable desks that combines style and comfort primarily for stationary positions making it ideal for sit-stand desks and standing workstations
  • A built-in handle makes it easy to carry or hang up when not in use
Specifications
Dimensions (L x W) 70 x 52 cm
Color Gray
Length (English) 2.3 ft.
Length (Metric) 70 cm
Material PVC
Type Industrial and Commercial Matting
Width (Metric) 52 cm