Learn More
Brady™ Post Top
This Post Top is ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
Brand:  Brady™ 292247
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11786675
  • 90.60 NOK / Each
Estimated Shipment: 29-07-2024
Description
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Top only.
Specifications
Specifications
1 Ea.
Black
Post Top
60 mm