Learn More
Brady™ Posts and Rubber Bases
Description
- Ideal for controlling and directing foot traffic.
- Suitable for warehouses, factories, offices etc.
- Chains and ribbons can be attached to the sturdy 900 mm posts.
- Post and Fillable Base.
Specifications
Specifications
| Alternate Part Number | POST & RUBBER BASE WHITE PK 4 |
| Diameter (Metric) Outer | 40 mm |
| Height (English) | 35.43 in. |
| Height (Metric) | 900 mm |
| Packaging Quantity | 4 Pk. |
| Color | White |
| Type | Post with Rubber Base |