Learn More
Brady™ Posts and Fillable Bases
Product Code. 11741306
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
Product Code. Color unitSize
11741306 White 1 pack
11701306 Red on White 1 pack
11711306 Black and Yellow Striped 1 pack
11761306 Red 1 pack
4 options
Product Code. 11741306
Brand: Brady™ 256725
Only null left
Add to basket
Add to basket
These Posts and Fillable Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Posts and Fillable Bases.
Specifications
Alternate Part Number POST & FILLABLE BASE WHITE PK 4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color White
Type Post with Fillable Base