Learn More
Brady™ Post and Cement Bases
Product Code. 11741316
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
Product Code. Color unitSize
11741316 White 1 pack
11721316 Red on White 1 pack
11791306 Red 1 pack
11711316 Black and Yellow Striped 1 pack
4 options
Product Code. 11741316
Brand: Brady™ 292239
Only null left
Add to basket
Add to basket
These Post and Cement Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Includes Post and Cement Base.
Specifications
Alternate Part Number BRADYPOST & CEMENT BASE WHITE PK4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color White
Type Post with Cement Base