Learn More
Smiths Medical Portex™ Fine Bore LDPE Tubing
Product Code. 15249134
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
0.38 mm
Unit Size:
30m
Product Code. Diameter (Metric) Inner unitSize
15249134 0.38 mm 30m
1 options
Product Code. 15249134
Brand: Smiths Medical 800/100/120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offering softness, a light weight and excellent chemical resistance. Smiths Medical™ Portex™ Fine Bore LDPE Tubing has a smooth bore for ease of cleaning.
  • Lightweight and flexible
  • Translucent in color
  • Surgical non-toxic quality, minimises trauma when used in vivo
  • Suitable for sheathing glass fibre in telecommunications
Specifications
Diameter (Metric) Inner 0.38 mm
Material LDPE
Diameter (Metric) Outer 1.09 mm
Overall Length (Metric) 30 m
For Use With (Application) For general laboratory applications
Min. Operating Temperature (Metric) -20°C
Packaging Quantity 1 Each
Color Translucent
Maximum Pressure 206 psi
Max. Operating Temperature (Metric) 80°C