missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Portable Waterproof pH/ORP Meter

Product Code. 10054300
Change view
Click to view available options
Type:
Portable Waterproof pH/ORP Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10054300 Portable Waterproof pH/ORP Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10054300 Supplier Hanna Instruments™ Supplier No. HI98150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Stores calibration data performed with a specific meter to provide information on electrode condition and performance, as well as date and time of last calibration. Hanna Instruments™ Portable Waterproof pH/ORP Meter is a compact and robust waterproofed meter complete with a SMART electrode.

  • Log-on-demand on board memory for up to 500 readings
  • Adjustable number of calibration points, from 2 to 5 points
  • Computer communication through USB
  • Backlit, multi level LCD display
  • Waterproof rubberised keyboard
  • Operational instructions on screen
  • Electrode incorporates temperature sensor for easier use
  • Automatic recognition of pH electrode including last date calibrated
  • High accuracy mV range for use with ORP/redox electrodes

Specifications

Includes Amplified pH electrode with built-in temperature sensor and EEPROM and DIN connector, HI70004 pH 4.01 sachet (1 pc), HI70007 pH 7.01 sachet (1 pc), Instruction Manual, AA size Alkaline Batteries (4 pcs), Rugged Carrying Case
Accuracy (Temperature) ±0.4°C from 0 to 70°C, ±1°C outside
Accuracy (mV) ±0.2 between ±400, ±0.4 from +400 to +600
Accuracy (pH) ±0.01
Outputs RS-232
Calibration Automatic
Range (pH) -4.00 to 19.99
Resolution (pH) 0.01
Range (mV) ±600, ±2000 Autoranging
Resolution (mV) 0.1 mV between ±400mV, 0.2 mV from +400 to +600mV, 0.2 mV from -400 to -600mV, 1 mV outside
Calibration Points 1 or 2 with 5 memorized buffers (4.01, 6.86, 7.01, 9.18 and 10.01 pH)
Temperature Range (Metric) -20.0°C to +120.0°C
Resolution (Temperature) 0.1°C from -10 to 120°C / 1°C below -10°C
Display Type LCD, Backlit
Memory Log up to 500 samples
Inputs DIN connector
Certifications/Compliance IP67
Dimensions (L x W x H) 196 x 80 x 60 mm (7.7 x 3.1 x 2.4 in.)
Weight (Metric) 500 g
Weight (English) 18 oz.
Warranty 2 Years (6 Months Probe)
Power Supply 1.5V AA batteries, 12 VDC adapter
Probe Type Amplified, with built-in EEPROM and DIN connector
Relative Humidity Range 1
Detection Range -2.00 to 19.99 pH
Temperature Compensation Automatic, –20 to 120°C
Temperature (Metric) Operating 0°C to 50°C
Battery Type 4 x 1.5V AA
Type Portable Waterproof pH/ORP Meters
Battery Life 300 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.