Learn More
OHAUS™ Portable Precision Balance
Product Code. 17217840 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
10,200 g
120 g
1200 g
220 g
2200 g
320 g
3200 g
Dimensions (L x W x H):
230 x 204 x 70 mm
250 x 240 x 74 mm
230 x 204 x 104 mm
Unit Size:
1 unit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Dimensions (L x W x H) Unit Size
17217840 120 g 230 x 204 x 104 mm 1 unit
17227840 220 g 230 x 204 x 104 mm 1 unit
17237840 320 g 230 x 204 x 104 mm 1 unit
17247840 1200 g 230 x 204 x 70 mm 1 unit
17257840 2200 g 230 x 204 x 70 mm 1 unit
17267840 3200 g 230 x 204 x 70 mm 1 unit
17277840 10,200 g 250 x 240 x 74 mm 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17217840 Supplier OHAUS™ Supplier No. 30615581
  • ONLINE EXCLUSIVE PRICE
    4345.00 NOK
    3162.00 NOK / Pack of 1
    Save 1183.00 NOK
    27% Off
View all promotions
 Restricted Item
In Stock
Multi-Purpose Portable Balances Suitable for Everyday Weighing
  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232, USB, or Ethernet (available as accessories)
Description Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Capacity 120 g
Display Type Backlit LCD
Readability 0.001 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Checkweighing
Linearity 0.003 g
Material ABS housing, stainless steel pan, transportation lock, slip-resistant and adjustable feet, leveling bubble
Model NV123
Repeatability 0.002 g
Stabilization Time 2 sec
Temperature Range (English) 50°F to 86°F, 80%RH, non-condensing¶operability guaranteed between 41°F to 104°F
Temperature Range (Metric) 10°C to 30°C, 80%RH, non-condensing¶operability guaranteed between 5°C to 40°C
Type Portable Balances
Dimensions (L x W x H) 230 x 204 x 104 mm
Length (English) 9.1 in.
Width (English) 8 in.
Height (English) 4.2 in.
Diameter (English) Outer Pan 3.7 in.
Diameter (Metric) Outer Pan 93 mm
Weight (English) 2.2 lb.