Learn More
Hanna Instruments™ ISO Compliant Portable Turbidity Meters
Descripción
- High accuracy benchtop turbidity meter used to determine the turbidity of water and wastewater with high precision in the field as well as in the laboratory
- The meter is supplied complete with AMCO-AEPA-1 primary turbidity standards used for calibration and performance verification
- Specially designed for water quality measurements, providing reliable and accurate readings, even within low turbidity ranges
- Based on a state-of-the-art optical system which guarantees accurate results, assures long term stability, and minimizes stray light and color interferences
- Periodic calibration with the supplied standards compensates for any variations in intensity of the LED light source
- The 25 mm round cuvettes composed of special optical glass guarantee the repeatability of turbidity measurements
- Meets and exceeds the meter criteria specied by ISO 7027
Especificaciones
Especificaciones
| Type | Portable Logging Turbidity Meter |
| Light Source | Infrared LED |
| Display Type | LED |
| Calibration Type | 3 points |
| Auto Shut Down Time | After 5 minutes of non-use |
| For Use With (Application) | Turbidity meters that provide laboratory accuracy for field turbidity measurements |
| Accuracy | ±5% |
| Resolution | 1FTU |
| Battery Life | Approximately 60 hours of continuous use or 900 measurements |
| Battery Type | 1.5V Battery |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?