missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Porcine IL-4, AOF Recombinant Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Animal Origin Free Reconstitution: Sterile Water at 0.1 mg/mL
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifications
Specifications
| Accession Number | Q04745 |
| For Use With (Application) | Control |
| Formulation | 10 mM sodium phosphate with no preservative; pH 7.5 |
| Gene ID (Entrez) | 397225 |
| Molecular Weight (g/mol) | 12.7 kDa |
| Name | Porcine IL-4, AOF |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | ≤0.1 EU/μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction