missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Porcine GM-CSF, AOF Recombinant Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Animal Origin Free Reconstitute with sterile water at 0.1 mg/mL.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
Specifications
Specifications
| Accession Number | Q29118 |
| Concentration | 0.1 mg/mL |
| For Use With (Application) | Control |
| Formulation | 10 mM sodium phosphate with no preservative |
| Gene ID (Entrez) | 397208 |
| Molecular Weight (g/mol) | 14.5 kDa |
| Name | Porcine GM-CSF, AOF |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction