missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Porcine GM-CSF, AOF Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87802
Additional Details : Weight : 0.01000kg
Description
GM-CSF (Granulocyte-Macrophage colony-stimulating factor) is a 14.6kDa hematopoietic growth factor that exists in glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The active form of the GM-CSF protein is found extracellularly as a homodimer and the GM-CSF gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q-syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. The ability of recombinant GM-CSF to increase hematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation. Recent studies have investigated GM-CSF in inflammatory and autoimmune diseases such as arthritis, arthritis related interstitial lung disease, nephritis, and psoriasis. In the CNS, GM-CSF depletion or neutralization has been studied in experimental autoimmune encephalomyelitis (EAE) and multiple sclerosis (MS).- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 128 aa.
- Biological Activity: <10 ng/mL
- Molecular Weight: 14.5 kDa
- Animal Origin Free: Yes
Specifications
Q29118 | |
10 mM sodium phosphate with no preservative | |
14.5 kDa | |
500 μg | |
-20°C | |
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); Colony-stimulating factor; CSF; Csf2; Csfgm; GMCSF; GM-CSF; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; Granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; Molgramostin; put. GM-CSF; Sargramostim | |
Porcine | |
Recombinant | |
>95% |
Control, ELISA, Functional Assay, Gel Shift, Western Blot | |
397208 | |
Porcine GM-CSF, AOF | |
E. coli | |
RUO | |
CSF2 | |
Unconjugated | |
Lyophilized |