Learn More
Haldenwanger™ Porcelain Tall Form Melting Crucibles
Product Code. 10201301
Change view
Click to view available options
Capacity (Metric):
15 mL
24 mL
85 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10201301 85 mL Each
10569601 15 mL Each
10323271 24 mL Each
3 options
Product Code. 10201301
Brand: Haldenwanger™ L 79C/4
Only null left
Add to basket
Add to basket
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Tall Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Capacity (Metric) 85 mL
Diameter (Metric) Outer 50mm
Height (Metric) 60mm
Product Type Melting Crucible