Learn More
Haldenwanger™ Porcelain Spouted Mortar
Product Code. 12373328
Click to view available options
Diameter (Metric) Outer:
65mm
130mm
150mm
180mm
250mm
330mm
Unit Size:
Each
Product Code. 12373328
Brand: Haldenwanger™ L 55/8/RAUH
Only null left
Add to basket
Add to basket
Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.
Grinding surfaces are unglazed.
Specifications
Capacity (Metric) 1000 mL
Diameter (Metric) Outer 180mm
Product Type Mortar
Material Porcelain
Height (Metric) 80mm
Includes With spout
Type Mortar