Learn More
Haldenwanger™ Porcelain Round Bottom Evaporating Basins
Product Code. 12035033
Click to view available options
Capacity (Metric):
180 mL
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12035033
Brand: Haldenwanger™ L 109/2
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Specifications
Material Porcelain
Capacity (Metric) 180 mL
Diameter (Metric) Outer 100mm
Height (Metric) 40mm
Product Type Evaporating Basin