Learn More
Haldenwanger™ Porcelain Mortars
Product Code. 11970897
Change view
Click to view available options
Capacity (Metric):
70 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11970897 70 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11970897 Supplier Haldenwanger™ Supplier No. L 55/0A/RAUH
  • ONLINE EXCLUSIVE PRICE
    273.00 NOK
    225.52 NOK / Pack of 1
    Save 47.48 NOK
    17% Off
Estimated Shipment: 28-12-2026
Manufactured from porcelain for high wear conditions. Haldenwanger™ Porcelain Mortars provide a rough surface for grinding materials to a fine powder.
  • Spout for easy pouring
  • Use with Haldenwanger™ pestles
  • Available in a variety of sizes to meet crushing, grinding and mixing needs in a laboratory
Specifications
Diameter (Metric) Inner 63mm
Product Type Mortar
Material Porcelain
Finish Rough Inner, Glazed Outer
Capacity (Metric) 70 mL