Learn More
Haldenwanger™ Porcelain Medium Form Crucibles
Product Code. 10539411
Change view
Click to view available options
Capacity (Metric):
10 mL
60 mL
150 mL
Unit Size:
Each
Pack of 10
Product Code. Capacity (Metric) unitSize
10539411 150 mL Each
12316507 150 mL Pack of 10
12336497 10 mL Pack of 10
12386497 60 mL Pack of 10
4 options
Product Code. 10539411
Brand: Haldenwanger™ L 79MF/2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications
Description Lid is sold separately; see 12316497 (79D/2).
Material Porcelain
Capacity (Metric) 150 mL
Product Type Crucible