Learn More
Haldenwanger™ Porcelain Incinerating Dishes
Product Code. 12085063
Change view
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
12085063 10 mL Each
1 options
Product Code. 12085063
Brand: Haldenwanger™ L 33D/4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Bottom Shape Flat
Capacity (Metric) 10 mL
Material Porcelain
Shape Round
Type Incinerating Dish
Height (Metric) 12 mm
Diameter (Metric) 47 mm