Learn More
Haldenwanger™ Porcelain Hirsch Funnel
Product Code. 12388567
Change view
Click to view available options
Diameter (Metric) Plate:
9mm
11mm
13mm
Unit Size:
Each
Height (Metric):
45 mm
56 mm
Product Code. Diameter (Metric) Plate unitSize Height (Metric)
12388567 13mm Each -
10717211 9mm Each 45 mm
10190131 11mm Each 56 mm
3 options
Product Code. 12388567
Brand: Haldenwanger™ L 126/4/0A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With perforated fixed filter plate
  • Available in a variety of sizes
  • With perforated fixed filter plate
  • Complies with DIN VDE 0335, type C 100
With perforated fixed filter plate.
Specifications
Material Porcelain
Diameter (Metric) Plate 13mm
Diameter (Metric) Disc 13mm
Includes With fixed filter plate
Quantity 1Each
Product Type Hirsch Funnel