Learn More
Haldenwanger™ Porcelain Hirsch Funnel
Product Code. 10190131
Click to view available options
Diameter (Metric) Plate:
9mm
11mm
13mm
Height (Metric):
45 mm
56 mm
Unit Size:
Each
Product Code. 10190131
Brand: Haldenwanger™ 126/4/0
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With perforated fixed filter plate
With perforated fixed filter plate.
Specifications
Material Porcelain
Height (Metric) 56 mm
Diameter (Metric) Plate 11mm
Diameter (Metric) Disc 11mm
Includes With fixed filter plate
Quantity 1Each
Product Type Hirsch Funnel