Learn More
Haldenwanger™ Porcelain Evaporating Dishes
Description
Sizes up to 175mL glazed except rim; 1,000mL and larger are glazed except outside base.
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 10 mL |
| Material | Porcelain |
| Shape | Round |
| Type | Evaporating Dish |
| For Use With (Application) | Laboratories |
| Interior Finish | Smooth |
| Height (Metric) | 11 mm |
| Diameter (Metric) | 50 mm |