Learn More
Haldenwanger™ Porcelain Desiccator Plates
Product Code. 10241481
Click to view available options
Diameter (Metric) Outer:
90 mm
140 mm
190 mm
235 mm
280 mm
Unit Size:
Each
Product Code. 10241481
Brand: Haldenwanger™ L 119C/280
Only null left
Add to basket
Add to basket
Use these plates for a variety of applications. Haldenwanger™ Porcelain Desiccator Plates feature a smooth internal finish, constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications
Diameter (Metric) Outer 280 mm
No. per Pack 1 Each
Product Type Desiccator Plate
Material Porcelain