Learn More
Haldenwanger™ Porcelain Crucible Lids
Product Code. 12316497
Change view
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
45mm
60mm
75mm
87mm
92mm
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
12316497 75mm Pack of 5
10311252 60mm Each
10210961 45mm Each
10250911 40mm Each
10557301 30mm Each
12807703 87mm Pack of 5
12817703 92mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12316497 Supplier Haldenwanger™ Supplier No. L 79D/2
  • ONLINE EXCLUSIVE PRICE
    752.00 NOK
    635.36 NOK / Pack of 5
    Save 116.64 NOK
    15% Off
Estimated Shipment: 14-12-2026
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 75mm
Product Type Crucible Lid