Learn More
Haldenwanger™ Porcelain Crucible Lids
Product Code. 10250911
Change view
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
45mm
60mm
75mm
87mm
92mm
Unit Size:
Each
Pack of 5
Product Code. Diameter (Metric) Outer unitSize
10250911 40mm Each
10311252 60mm Each
10210961 45mm Each
10557301 30mm Each
12807703 87mm Pack of 5
12817703 92mm Pack of 5
12316497 75mm Pack of 5
7 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 10250911
Brand: Haldenwanger™ L 79D/7A
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 40mm
Product Type Crucible Lid