Learn More
Haldenwanger™ Porcelain Crucible Lids
Product Code. 10210961
Change view
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
45mm
60mm
75mm
87mm
92mm
Unit Size:
Each
Pack of 5
Product Code. Diameter (Metric) Outer unitSize
10210961 45mm Each
10311252 60mm Each
10250911 40mm Each
10557301 30mm Each
12807703 87mm Pack of 5
12817703 92mm Pack of 5
12316497 75mm Pack of 5
7 options
Product Code. 10210961
Brand: Haldenwanger™ L 79D/6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 45mm
Product Type Crucible Lid