Learn More
Haldenwanger™ Porcelain Casseroles
Product Code. 10701392
Change view
Click to view available options
Capacity (Metric):
140 mL
220 mL
500 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10701392 140 mL Each
10751562 500 mL Each
10220961 220 mL Each
3 options
Product Code. 10701392
Brand: Haldenwanger™ 17/000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Capacity (Metric) 140 mL
Material Porcelain
Shape Round
Type Casserole
Height (Metric) 35 mm
Diameter (Metric) 63 mm