Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Product Code. p-8008540
Change view
Click to view available options
Capacity (Metric):
660 mL
Diameter (Metric) Plate:
125mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Plate unitSize
11737335 660 mL 125mm Each
1 options
Product Code. 11737335
Brand: Rogo-Sampaic PNF013
Only null left
Add to basket
Add to basket
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
  • White porcelain
  • Available for a range of volumes and filter diameters
  • Applications include collecting recrystalized compounds
  • Excellent thermal and chemical resistance
Specifications
Capacity (Metric) 660 mL
Material Porcelain
Diameter (Metric) Plate 125mm
Diameter (Metric) Disc 125mm
Length (Metric) Stem 85mm
Quantity 1Each
Product Type Buchner Funnel