Learn More
Haldenwanger™ Porcelain Basins
Product Code. 10048160
Change view
Click to view available options
Height (Metric):
10 mm
13 mm
15 mm
16 mm
Length (Metric):
45 mm
50 mm
70 mm
96 mm
Unit Size:
Each
Product Code. Height (Metric) Length (Metric) unitSize
10048160 13 mm 70 mm Each
10509031 10 mm 45 mm Each
10260961 15 mm 50 mm Each
10038150 16 mm 96 mm Each
4 options
Product Code. 10048160
Brand: Haldenwanger™ L 33C/4B
Only null left
Add to basket
Add to basket
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
  • Shallow form porcelain basin
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Shape Rectangular
Type Evaporating Basin
For Use With (Application) Laboratories
Interior Finish Smooth
Dimensions (L x W x H) 70 x 46 x 13 mm
Length (Metric) 70 mm
Height (Metric) 13 mm